Merck
  • PKB/Akt: a key mediator of cell proliferation, survival and insulin responses?
PKB/Akt: a key mediator of cell proliferation, survival and insulin responses?
Journal of cell science (2001-11-01)
M A Lawlor, D R Alessi
ABSTRACT
The serine/threonine protein kinase PKB (also known as Akt) is thought to be a key mediator of signal transduction processes. The identification of PKB substrates and the role PKB phosphorylation plays in regulating these molecules have been a major focus of research in recent years. A recently developed motif-profile scoring algorithm that can be used to scan the genome for potential PKB substrates is therefore a useful tool, although additional considerations, such as the evolutionary conservation of the phosphorylation site, must also be taken into account. Recent evidence indicates that PKB plays a key role in cancer progression by stimulating cell proliferation and inhibiting apoptosis and is also probably a key mediator of insulin signalling. These findings indicate that PKB is likely to be a hot drug target for the treatment of cancer, diabetes and stroke. There are, however, a number of pitfalls of methodologies currently employed to study PKB function, and therefore caution should be used in interpretation of such experiments.