Merck
  • Standing wave-assisted acoustic droplet vaporization for single and dual payload release in acoustically-responsive scaffolds.
Standing wave-assisted acoustic droplet vaporization for single and dual payload release in acoustically-responsive scaffolds.
Ultrasonics sonochemistry (2020-04-06)
Mitra Aliabouzar, Aniket Jivani, Xiaofang Lu, Oliver D Kripfgans, J Brian Fowlkes, Mario L Fabiilli
ABSTRACT
An ultrasound standing wave field (SWF) has been utilized in many biomedical applications. Here, we demonstrate how a SWF can enhance drug release using acoustic droplet vaporization (ADV) in an acoustically-responsive scaffold (ARS). ARSs are composite fibrin hydrogels containing payload-carrying, monodispersed perfluorocarbon (PFC) emulsions and have been used to stimulate regenerative processes such as angiogenesis. Elevated amplitudes in the SWF significantly enhanced payload release from ARSs containing dextran-loaded emulsions (nominal diameter: 6 μm) compared to the -SWF condition, both at sub- and suprathreshold excitation pressures. At 2.5 MHz and 4 MPa peak rarefactional pressure, the cumulative percentage of payload released from ARSs reached 84.1 ± 5.4% and 66.1 ± 4.4% under + SWF and -SWF conditions, respectively, on day 10. A strategy for generating a SWF for an in situ ARS is also presented. For dual-payload release studies, bi-layer ARSs containing a different payload within each layer were exposed to temporally staggered ADV at 3.25 MHz (day 0) and 8.6 MHz (day 4). Sequential payload release was demonstrated using dextran payloads as well as two growth factors relevant to angiogenesis: basic fibroblast growth factor (bFGF) and platelet-derived growth factor BB (PDGF-BB). In addition, bubble growth and fibrin degradation were characterized in the ARSs under +SWF and -SWF conditions. These results highlight the utility of a SWF for modulating single and dual payload release from an ARS and can be used in future therapeutic studies.
Sigma-Aldrich
Fibroblast Growth Factor basic, human recombinant, animal-free, The Fibroblast Growth Factor-basic (or bFGF protein) is a heparin binding growth factor which stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal & endothelial cells.
Pricing and availability is not currently available.