Merck
  • Identification and Quantitation of Hydroxy Fatty Acids in Fermented Sausage Samples.
Identification and Quantitation of Hydroxy Fatty Acids in Fermented Sausage Samples.
Journal of agricultural and food chemistry (2020-07-17)
Nuanyi Liang, Kaixing Tang, Jonathan M Curtis, Michael G Gänzle
ABSTRACT
The quality of fermented sausage is strongly influenced by its fatty acid (FA). However, the role of a defined starter culture in modifying sausage FA composition, and especially in the production of hydroxy FAs (HFAs), has not been determined. In this study, the FA compositions of sausages fermented with Latilactobacillus sakei, with L. sakei plus Staphylococcus carnosus, and with an aseptic control were characterized by liquid chromatography-mass spectrometry (MS)/MS and gas chromatography-MS. The sausages fermented with L. sakei, and with L. sakei plus S. carnosus, showed a reduced accumulation of poly and/or diunsaturated FAs and distinct composition of HFAs compared to the aseptic control. 2-HFAs were enriched via high-speed counter-current chromatography and identified uniquely in the L. sakei plus S. carnosus fermented sausage. Through lipid analyses, this study illustrated how the choice of a defined starter culture affected the observed FA metabolism in fermented sausages, facilitating the development of starter cultures or additives that impart desirable characteristics to sausage.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Boron trifluoride-methanol solution, 14% in methanol
Sigma-Aldrich
2OHOA, ≥98% (HPLC)