Merck
  • Use of internally validated in vitro biofilm models to assess antibiofilm performance of silver-containing gelling fibre dressings.
Use of internally validated in vitro biofilm models to assess antibiofilm performance of silver-containing gelling fibre dressings.
Journal of wound care (2020-03-12)
Louise Suleman, Liam Purcell, Hannah Thomas, Samantha Westgate
ABSTRACT
To assess the efficacy of five silver-containing gelling fibre wound dressings against single-species and multispecies biofilms using internally validated, UKAS-accredited in vitro test models. Staphylococcus aureus, Pseudomonas aeruginosa and Candida albicans single- and multispecies biofilms were cultured using Centres for Disease Control (CDC) biofilm reactors and colony drip flow reactors (CDFR). Following a 72 hour incubation period, the substrates on which biofilms were grown were rinsed to remove planktonic microorganisms and then challenged with fully hydrated silver-containing gelling fibre wound dressings. Following dressing application for 24 or 72 hours, remaining viable organisms from the treated biofilms were quantified. In single-species in vitro models, all five antimicrobial dressings were effective in eradicating Staphylococcus aureus and Pseudomonas aeruginosa biofilm bacteria. However, only one of the five dressings (Hydrofiber technology with combination antibiofilm/antimicrobial technology) was able to eradicate the more tolerant single-species Candida albicans biofilm. In a more complex and stringent CDFR biofilm model, the hydrofiber dressing with combined antibiofilm/antimicrobial technology was the only dressing that was able to eradicate multispecies biofilms such that no viable organisms were recovered. Given the detrimental effects of biofilm on wound healing, stringent in vitro biofilm models are increasingly required to investigate the efficacy of antimicrobial dressings. Using accredited in vitro biofilm models of increasing complexity, differentiation in the performance of dressings with combined antibiofilm/antimicrobial technology against those with antimicrobial properties alone, was demonstrated.
Product Description
Millipore
Universal Microbial Limit Test Membrane, white colored, sterile, diameter 70 mm, suitable for microbiology