- ADAM12-mediated focal adhesion formation is differently regulated by beta1 and beta3 integrins.
ADAM12-mediated focal adhesion formation is differently regulated by beta1 and beta3 integrins.
FEBS letters (2005-10-11)
Charles Kumar Thodeti, Camilla Fröhlich, Christian Kamp Nielsen, Yoshikazu Takada, Reinhard Fässler, Reidar Albrechtsen, Ulla M Wewer
PMID16213489
ABSTRACT
ADAM12, adisintegrin and metalloprotease, has been demonstrated to be upregulated in human malignant tumors and to accelerate the malignant phenotype in a mouse model for breast cancer. ADAM12 is a substrate for beta1 integrins and may affect tumor and stromal cell behavior through its binding to beta1 integrins. Here, we report that cells deficient in beta1 integrin or overexpressing beta3 integrin can bind to recombinant full-length human ADAM12 via beta3 integrin. Furthermore, cell binding to ADAM12 via beta3 integrin results in the formation of focal adhesions, which are not formed upon beta1 integrin-mediated cell attachment. We also show that RhoA is involved in beta3 integrin-mediated focal adhesion formation.
Sigma-Aldrich
Anti-Integrin Beta1, clone AIIB2 (Azide Free) Antibody, clone AIIB2, 1 mg/mL, from rat