Merck
  • Regulation of the unfolded protein response in yeast by oxidative stress.
Regulation of the unfolded protein response in yeast by oxidative stress.
FEBS letters (2019-04-20)
Angel Guerra-Moreno, Jessie Ang, Hendrik Welsch, Marco Jochem, John Hanna
ABSTRACT
In the unfolded protein response (UPR), Ire1 activates Hac1 to coordinate the transcription of hundreds of genes to mitigate ER stress. Recent work in Caenorhabditis elegans suggests that oxidative stress inhibits this canonical Ire1 signalling pathway, activating instead an antioxidant stress response. We sought to determine whether this novel mode of UPR function also existed in yeast, where Ire1 has been best characterized. We show that the yeast UPR is also subject to inhibition by oxidative stress. Inhibition is mediated by a single evolutionarily conserved cysteine, and affects both luminal and membrane pathways of Ire1 activation. In yeast, Ire1 appears dispensable for resistance to oxidative stress and, therefore, the physiological significance of this pathway remains to be demonstrated.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.