Merck
  • Novel bisphosphonate inhibitors of the human farnesyl pyrophosphate synthase.
Novel bisphosphonate inhibitors of the human farnesyl pyrophosphate synthase.
Bioorganic & medicinal chemistry letters (2010-08-31)
Joris W De Schutter, Serge Zaretsky, Sarah Welbourn, Arnim Pause, Youla S Tsantrizos
ABSTRACT
A structure-based approach was pursued in designing novel bisphosphonate inhibitors of the human farnesyl pyrophosphate synthase (hFPPS). Preliminary SAR and structural evidence for the simultaneous binding of these inhibitors into the isopentenyl pyrophosphate (IPP) and the geranyl pyrophosphate (GPP) substrate sub-pockets of the enzyme are presented.
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Geranyl pyrophosphate lithium salt, analytical standard