Merck
  • Knockdown of Bone Morphogenetic Proteins Type 1a Receptor (BMPR1a) in Breast Cancer Cells Protects Bone from Breast Cancer-Induced Osteolysis by Suppressing RANKL Expression.
Knockdown of Bone Morphogenetic Proteins Type 1a Receptor (BMPR1a) in Breast Cancer Cells Protects Bone from Breast Cancer-Induced Osteolysis by Suppressing RANKL Expression.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-03-02)
Yang Liu, Ran-Xi Zhang, Wei Yuan, Hou-Qing Chen, Dong-Dong Tian, Hao Li, Xiao Jiang, Zhong-Liang Deng, Yang Wang
ABSTRACT
Bone morphogenetic proteins (BMPs) and BMP receptors widely participate in osteolytic metastasis of breast cancer, while their role in tumor-stromal interaction is largely unknown. In this study, we investigated whether BMP receptor type 1a (BMPR1a) can alter the interaction between metastatic cancer cells and osteoclast precursors. Adenovirus-mediated RNA interference was used to interrupt target genes of human breast cancer cell lines and nude mice were injected intratibially with the cancer cells. Tumor-bearing mice were examined by bioluminescence imaging and microCT. Sections of metastatic legs were measured by a series of staining methods. Murine bone marrow mononuclear cells or RAW264.7 cells were cultured with conditioned media of breast cancer cells. RT-PCR, Western blotting and ELISA were used to test mRNA and protein expressions of target molecules. Expression of BMPR1a of MDA-MB-231-luc cells at tumor-bone interface was apparently stronger than that of cancer cells distant from the interface. Mice injected with BMPR1a-knockdown MDA-MB-231-luc cells showed reduced tumor growth and bone destruction compared with control groups. Knockdown (KD) of BMPR1a of MDA-MB-231-luc cells or MCF-7 cells decreased the level of receptor activator for NF-κB ligand (RANKL). Level of RANKL in MDA-MB-231-luc cells or MCF-7 cells was reduced by p38 inhibitor. Compared with control group, knockdown of p38 of breast cancer cells decreased cancer-induced osteoclastogenesis. Knockdown of BMPR1a of breast cancer cells suppresses their production of RANKL via p38 pathway and inhibits cancer-induced osteoclastogenesis, which indicates that BMPR1a might be a possible target in breast cancer-induced osteolytic metastasis.
Sigma-Aldrich
Anti-TNFSF11 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Osteoprotegerin antibody produced in goat, affinity isolated antibody, lyophilized powder
Pricing and availability is not currently available.