
Trimethadione

Synonym(s):
3,5,5,-Trimethyloxazolidine-2,4-dione, Trimethadione
Empirical Formula (Hill Notation):
C6H9NO3
CAS Number:
Molecular Weight:
143.14
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing