
Triamcinolone acetonide for system suitability

Synonym(s):
Triamcinolone acetonide, 9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide
Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing