
Spinosin

Synonym(s):
Flavoayamenin
Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing