
Plastic additive 24

Synonym(s):
Diisononyl cyclohexane-1,2-dicarboxylate
Empirical Formula (Hill Notation):
C26H48O4
CAS Number:
Molecular Weight:
424.66
Products
Formula Weight
Manufacturer
Application
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing