
Plastic additive 17

Synonym(s):
Distearyl 3,3′-thiodipropionate, 3,3′-Thiodipropionic acid di-n-octadecyl ester, Dioctadecyl 3,3′-thiodipropionate, Thiodipropionic acid distearyl ester
Linear Formula:
S[CH2CH2CO2(CH2)17CH3]2
CAS Number:
Molecular Weight:
683.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing