
Plastic additive 10

Synonym(s):
1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene
Linear Formula:
(CH3)3C6[CH2C6H2[C(CH3)3]2OH]3
CAS Number:
Molecular Weight:
775.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing