
Phospholipase C from Bacillus cereus

Synonym(s):
Lecithinase C, Lipophosphodiesterase I, PC-PLC, Phosphatidylcholine cholinephosphohydrolase
CAS Number:
Products
Brand
Sustainability
Feature
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing