
Noscapine

Synonym(s):
(S,R)-Noscapine
Empirical Formula (Hill Notation):
C22H23NO7
CAS Number:
Molecular Weight:
413.42
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing