
Norhydrocodone-d3 hydrochloride

Synonym(s):
Norhydrocodone-D3 hydrochloride solution
Empirical Formula (Hill Notation):
C17D3H16NO3 · HCl
Molecular Weight:
324.82
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing