Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse
Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing