
Flecainide for system suitability

Synonym(s):
Flecainide acetate salt, N-(2-Piperidylmethyl)-2,5-bis-(2,2,2-trifluoroethoxy)benzamide acetate salt
Empirical Formula (Hill Notation):
C17H20F6N2O3 · C2H4O2
CAS Number:
Molecular Weight:
474.39
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing