
Flavoxate hydrochloride

Synonym(s):
3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid 2-(1-piperidinyl)ethyl ester hydrochloride, DW-61, Rec-7-0040
Empirical Formula (Hill Notation):
C24H25NO4 · HCl
CAS Number:
Molecular Weight:
427.92
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing