
Doxepin Related Compound C

Synonym(s):
Nordoxepin hydrochloride, 11(6H)-(3-[Methylamino]propylidene)dibenz[b,e]oxepine
Empirical Formula (Hill Notation):
C18H19NO · HCl
CAS Number:
Molecular Weight:
301.81
Products
Brand
Formula Weight
Physical Form
Agency
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing