Skip to Content
Merck
CDP-Star® Disodium 2-chloro-5-(4-methoxyspiro {1,2-dioxetane-3,2′-(5′-chloro)tricyclo[3.3.1.13,7 ]decan}-4-yl)-1-phenyl phosphate

CDP-Star®

CDP-Star® Disodium 2-chloro-5-(4-methoxyspiro {1,2-dioxetane-3,2′-(5′-chloro)tricyclo[3.3.1.13,7 ]decan}-4-yl)-1-phenyl phosphate
Synonym(s):
CDP-Star® Chemiluminescent Substrate, Disodium 2-chloro-5-(4-methoxyspiro[1,2-dioxetane-3,2′-(5-chlorotricyclo[3.3.1.13.7]decan])-4-yl]-1-phenyl phosphate
Empirical Formula (Hill Notation):
C18H19Cl2O7Na2P
CAS Number:
Molecular Weight:
495.20

Products

Formula Weight
Manufacturer
Physical Form
Quality Segment
Package Size