![BTT Activator 0.3 M](/deepweb/assets/sigmaaldrich/product/structures/421/773/98333e3c-4f5d-4ed2-9de1-aafa976f98c3/640/98333e3c-4f5d-4ed2-9de1-aafa976f98c3.png)
BTT Activator
![BTT Activator 0.3 M](/deepweb/assets/sigmaaldrich/product/structures/421/773/98333e3c-4f5d-4ed2-9de1-aafa976f98c3/640/98333e3c-4f5d-4ed2-9de1-aafa976f98c3.png)
Synonym(s):
5-(Benzylthio)-1H-tetrazole solution
Empirical Formula (Hill Notation):
C8H8N4S
CAS Number:
Molecular Weight:
192.24
Products
Product Category
Brand
Color
Formula Weight
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing