![BSTFA (with 1% TMCS) kit of 10 × 1 mL, analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/149/727/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d/640/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d.png)
BSTFA
![BSTFA (with 1% TMCS) kit of 10 × 1 mL, analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/149/727/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d/640/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d.png)
Synonym(s):
N,O-Bis(trimethylsilyl)trifluoroacetamide, BSTFA
Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing