![β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin ≥99%, lyophilized powder](/deepweb/assets/sigmaaldrich/product/structures/397/795/0cc8dbd0-e86a-4bae-8e77-ce5282b02aef/640/0cc8dbd0-e86a-4bae-8e77-ce5282b02aef.png)
β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin
![β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin ≥99%, lyophilized powder](/deepweb/assets/sigmaaldrich/product/structures/397/795/0cc8dbd0-e86a-4bae-8e77-ce5282b02aef/640/0cc8dbd0-e86a-4bae-8e77-ce5282b02aef.png)
Synonym(s):
1-O-Alkyl-2-acetyl-sn-glycero-3-phosphocholine, 2-Acetyl-1-alkyl-sn-glycero-3-phosphocholine, AcGEPC, PAF, Platelet activating factor
CAS Number:
Products
Product Category
Brand
Physical Form
Purity
Research Area
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing