![Anti-HA antibody, Mouse monoclonal clone HA-7, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/261/335/1ecf02f3-4387-4086-a40c-35d3c4ec2cf2/640/1ecf02f3-4387-4086-a40c-35d3c4ec2cf2.jpg)
Anti-HA antibody, Mouse monoclonal
![Anti-HA antibody, Mouse monoclonal clone HA-7, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/261/335/1ecf02f3-4387-4086-a40c-35d3c4ec2cf2/640/1ecf02f3-4387-4086-a40c-35d3c4ec2cf2.jpg)
Synonym(s):
Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing