![2-Dechloro Aripiprazole Pharmaceutical Analytical Impurity (PAI)](/deepweb/assets/sigmaaldrich/product/images/751/396/144e21f7-629c-4347-ae03-573d0b7e4696/640/144e21f7-629c-4347-ae03-573d0b7e4696.jpg)
2-Dechloro Aripiprazole
![2-Dechloro Aripiprazole Pharmaceutical Analytical Impurity (PAI)](/deepweb/assets/sigmaaldrich/product/images/751/396/144e21f7-629c-4347-ae03-573d0b7e4696/640/144e21f7-629c-4347-ae03-573d0b7e4696.jpg)
Synonym(s):
7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, 2-Deschloro-aripiprazole
Empirical Formula (Hill Notation):
C23H28ClN3O2
CAS Number:
Molecular Weight:
413.94
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing