Skip to Content
Merck
All Photos(1)

Documents

GE17-5111-01

HiTrap® IGY Purification

Cytiva 17-5111-01, pack of 1 × 5 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

2-mercaptopyridine

packaging

pack of 1 × 5 mL

manufacturer/tradename

Cytiva 17-5111-01

storage condition

(20% Ethanol)

parameter

<20 mL/min flow rate
42 psi

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-13

working range

3-11

storage temp.

2-30°C

General description

HiTrap® IgY Purification HP is a 5 mL column prepacked with Sepharose High Performance for fast and easy purification of IgY from egg yolk.

Application

HiTrap® IgY Purification HP is packed with a thiophilic adsorption medium, 2-merCaptopyridine coupled to Sepharose High Performance. Thiophilic adsorption is promoted by water-structuring salts. It has been suggested that the interaction of IgY and ligand results from combined electron donating and accepting action of the ligand or, alternatively, a mixed mode hydrophilic-hydrophobic interaction.

Features and Benefits

  • Each column binds IgY from a one-quarter portion of egg yolk (approx. 100 mg IgY). For greater capacity, several columns may be connected in series.
  • Packed with a thiophilic adsorption medium, the ligand, 2-merCaptopyridine is coupled to Sepharose High Performance.
  • Recovery and purity of IgY are excellent.
  • Can be operated with a liquid chromatography system such as ÄKTA® design.
  • The top and bottom frits are manufactured from porous polyethylene
  • Connectors for usage with different chromatography systems and other equipment are included

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service