HPLC Analysis of Aspartic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec CLC-L, 15 cm x 4.6 mm I.D., 5 μm particles (53123AST)
mobile phase
5 mM copper sulfate
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
5 mg/mL in water