Merck
  • Triacylglycerol structure and interesterification of palmitic and stearic acid-rich fats: an overview and implications for cardiovascular disease.
Triacylglycerol structure and interesterification of palmitic and stearic acid-rich fats: an overview and implications for cardiovascular disease.
Nutrition research reviews (2009-05-16)
Sarah E E Berry
ABSTRACT
The position of fatty acids in the TAG molecule (sn-1, sn-2 and sn-3) determines the physical properties of the fat, which affects its absorption, metabolism and distribution into tissues, which may have implications for the risk of CHD. The TAG structure of fats can be manipulated by the process of interesterification, which is of increasing commercial importance, as it can be used to change the physical characteristics of a fat without the generation of trans-fatty acids. Interesterified fats rich in long-chain SFA are commercially important, but few studies have investigated their health effects. Evidence from animal and human infant studies suggests that TAG structure and interesterification affect digestibility, atherogenicity and fasting lipid levels, with fats containing palmitic and stearic acid in the sn-2 position being better digested and considered to be more atherogenic. However, chronic studies in human adults suggest that TAG structure has no effect on digestibility or fasting lipids. The postprandial effects of fats with differing TAG structure are better characterised but the evidence is inconclusive; it is probable that differences in the physical characteristics of fats resulting from interesterification and changes in TAG structure are key determinants of the level of postprandial lipaemia, rather than the position of fatty acids in the TAG. The present review gives an overview of TAG structure and interesterified palmitic and stearic acid-rich fats, their physical properties and their acute and chronic effects in human adults in relation to CHD.
Product Description
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard