Merck
  • Comparison of tolerability and efficacy of a combination of paracetamol + caffeine and sumatriptan in the treatment of migraine attack: a randomized, double-blind, double-dummy, cross-over study.
Comparison of tolerability and efficacy of a combination of paracetamol + caffeine and sumatriptan in the treatment of migraine attack: a randomized, double-blind, double-dummy, cross-over study.
The journal of headache and pain (2012-10-12)
Luigi Alberto Pini, Simona Guerzoni, Michela Cainazzo, Michela Ciccarese, Maria Pia Prudenzano, Paolo Livrea
ABSTRACT
In this study, we compared the efficacy and tolerability of the combination of paracetamol 1,000 mg + caffeine 130 mg (PCF) with sumatriptan 50 mg (SUM) in migraine attacks. This was a multi-center randomized double-blind, double-dummy, cross-over controlled trial. The efficacy was assessed by the sum of pain intensity differences, the curve of mean pain intensity, the number of pain free at 2 h, and the total pain relief. Tolerability was assessed by recording adverse events within 4 h after drug assumption and evaluating the global judgement of patients. The comparison of these parameters did not show differences between the two drugs which resulted absolutely overlapping in pain relief and patients evaluation. In conclusion, we confirm the efficacy and safety of PCF such as SUM in the treatment of migraine attacks.
Sigma-Aldrich
Sumatriptan succinate, ≥98% (HPLC), solid
Sumatriptan succinate, European Pharmacopoeia (EP) Reference Standard
Sumatriptan for system suitability, European Pharmacopoeia (EP) Reference Standard