Merck
  • Lupeol protects against acetaminophen-induced oxidative stress and cell death in rat primary hepatocytes.
Lupeol protects against acetaminophen-induced oxidative stress and cell death in rat primary hepatocytes.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-03-13)
Archana Kumari, Poonam Kakkar
ABSTRACT
Drug induced hepatotoxicity is a major problem where phytochemicals hold promise for its abrogation. This study was carried out to explore cytoprotective potential of lupeol, a triterpene, against acetaminophen (AAP)-induced toxicity in rat hepatocytes. AAP exposure significantly (p<0.05) reduced cell viability, disturbed Bcl-2 family pro/anti-apoptotic protein balance, increased ROS production and altered redox homeostasis. It also induced mitochondria-mediated hepatocellular injury by significant mitochondrial depolarization, caspase-9/3 activation and subsequent DNA fragmentation. Our results suggest that lupeol pre-treatment effectively restored antioxidant enzyme levels, decreased lipid peroxidation, inhibited ROS generation and depolarization of mitochondria. Lupeol also attenuated mitochondria-mediated signaling pathway and DNA damage as evident from TUNEL assay and cell cycle studies leading to prevention of cytotoxicity. This study confirms the efficacy of lupeol, a food derived antioxidant, in abrogating ROS generation, maintaining redox balance and providing significant protection against mitochondria-mediated cell death during AAP-induced toxicity.
Sigma-Aldrich
Lupeol, ≥94%
Supelco
Lupeol, analytical standard