Merck
  • TO901317 regulating apolipoprotein M expression mediates via the farnesoid X receptor pathway in Caco-2 cells.
TO901317 regulating apolipoprotein M expression mediates via the farnesoid X receptor pathway in Caco-2 cells.
Lipids in health and disease (2011-11-08)
Chunhua Zhu, Dongmei Di, Xiaoying Zhang, Guanghua Luo, Zongchun Wang, Jiang Wei, Yuanping Shi, Maria Berggren-Söderlund, Peter Nilsson-Ehle, Ning Xu
ABSTRACT
Apolipoprotein M (apoM) may have potential antiatherosclerotic properties. It has been reported that apoM expression could be regulated by many intracellar and extracellar factors. In the present study we further investigated regulation of apoM expression in Caco-2 cells stimulated by a liver X receptor (LXR) agonist, TO901317. Caco-2 cells were cultured in the presence of either TO901317, farnesoid X receptor (FXR) antagonist guggulsterone or TO901317 together with guggulsterone at different concentrations for 24 hrs. The mRNA levels of ATP-binding cassette transporter A1 (ABCA1), apoA1, apoM, liver receptor homologue-1 (LRH-1) and short heterodimer partner 1 (SHP1) were determined by real-time RT-PCR. When Caco-2 cell cultured with TO901317 alone, the mRNA levels of ABCA1, apoA1, apoM, LRH-1 and SHP1 were significantly increased with dose-dependent manners (p < 0.05), whereas when the cells cultured with guggulsterone alone, the mRNA levels of apoM, SHP1 and LRH-1 (p < 0.05) were strongly inhibited. Moreover, guggulsterone could abolish the TO901317 enhanced mRNA levels of apoA1 apoM, SHP1 and LRH-1. The present study demonstrated that LXR agonist TO901317 induced apoM expression in Caco-2 cells might be mediated via the LXR/FXR pathway.
Sigma-Aldrich
(E)-Guggulsterone, ≥95% (HPLC), powder
Sigma-Aldrich
(Z)-Guggulsterone, ≥89% (HPLC), powder