Merck
ProNGF is a potential diagnostic biomarker for thyroid cancer.
Oncotarget (2016-04-14)
Sam Faulkner, Severine Roselli, Yohann Demont, Jay Pundavela, Genevieve Choquet, Philippe Leissner, Christopher Oldmeadow, John Attia, Marjorie M Walker, Hubert Hondermarck
ABSTRACT
The precursor for nerve growth factor (proNGF) is expressed in some cancers but its clinicopathological significance is unclear. The present study aimed to define the clinicopathological significance of proNGF in thyroid cancer. ProNGF expression was analysed by immunohistochemistry in two cohorts of cancer versus benign tumors (adenoma) and normal thyroid tissues. In the first cohort (40 thyroid cancers, 40 thyroid adenomas and 80 normal thyroid tissues), proNGF was found overexpressed in cancers compared to adenomas and normal samples (p<0.0001). The area under the receiver-operating characteristic (ROC) curve was 0.84 (95% CI 0.75-0.93, p<0.0001) for cancers versus adenomas, and 0.99 (95% CI 0.98-1.00, p<0.0001) for cancers versus normal tissues. ProNGF overexpression was confirmed in a second cohort (127 cancers of various histological types and 55 normal thyroid tissues) and using a different antibody (p<0.0001). ProNGF staining intensity was highest in papillary carcinomas compared to other histological types (p<0.0001) and there was no significant association with age, gender, tumor size, stage and lymph node status. In conclusion, proNGF is increased in thyroid cancer and should be considered as a new potential diagnostic biomarker.
Sigma-Aldrich
Anti-Nerve Growth Factor Antibody, pro, serum, ChemiconĀ®