
TSKgel® UP-SW2000 Direct Connect (2 µm) HPLC Columns

Synonym(s):
TSKgel® UP-SW2000
Products
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing