
Tetrahydrozoline hydrochloride

Synonym(s):
4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole monohydrochloride, Tetryzoline hydrochloride, Tetrahydrozoline hydrochloride
Empirical Formula (Hill Notation):
C13H16N2 · HCl
CAS Number:
Molecular Weight:
236.74
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing