
Teriparatide for system suitability

Synonym(s):
Parathyroid Hormone Fragment 1-34 human, PTH 1-34, Parathormone (1-34)
Empirical Formula (Hill Notation):
C181H291N55O51S2
CAS Number:
Molecular Weight:
4117.72
Products
Formula Weight
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing