
Tenoxicam degradation impurity standard

Synonym(s):
Tenoxicam, 4-Hydroxy-2-methyl-N-2-pyridinyl-2H-thieno(2,3-e)-1,2-thiazine-3-carboxamide 1,1-dioxide
Empirical Formula (Hill Notation):
C13H11N3O4S2
CAS Number:
Molecular Weight:
337.37
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing