
Sulpiride impurity B

Synonym(s):
Methyl 2-methoxy-5-sulfamoylbenzoate
Linear Formula:
CH3OC6H3(SO2NH2)CO2CH3
CAS Number:
Molecular Weight:
245.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing