
Stanozolol impurity A

Synonym(s):
17α-Methylandrostan-17β-ol-3-one, 17α-Methyldihydrotestosterone, 17β-Hydroxy-17α-methyl-5α-androstan-3-one, 5α-Androstane-17α-methyl-17β-ol-3-one, Mestaline, Mestanolone, Methylandrostanolone
Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing