
Prednicarbate for peak identification

Synonym(s):
Prednicarbate, (11β)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C27H36O8
CAS Number:
Molecular Weight:
488.57
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing