
Poly(ethylene glycol) bis(amine)

Synonym(s):
O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)
Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
Products
Brand
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-6 of 6
Compare
Product Number
Description
Pricing