
Phenmedipham

Empirical Formula (Hill Notation):
C16H16N2O4
CAS Number:
Molecular Weight:
300.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2