
N-Nitroso-diisopropylamine

Synonym(s):
N-(1-Methylethyl)-N-nitroso-2-propanamine solution, N-Isopropyl-N-nitrosoisopropylamine, DIPNA, NDIP
Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing