
Nevirapine Related Compound A

Synonym(s):
11-Ethyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one
Empirical Formula (Hill Notation):
C14H14N4O
CAS Number:
Molecular Weight:
254.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing