
Methotrexate Related Compound E

Synonym(s):
4-{[(2,4-Diaminopteridin-6-yl)methyl](methyl)amino}benzoic acid hemihydrochloride
Empirical Formula (Hill Notation):
C15H15N7O2 · 0.5HCl
CAS Number:
Molecular Weight:
343.56
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2