
Magnetic iron oxide nanoparticles solution

Synonym(s):
Iron oxide(II,III), magnetic nanoparticles solution, Magnetic iron oxide nanocrystals, Magnetite, Superparamagnetic iron oxide nanoparticles
Empirical Formula (Hill Notation):
Fe3O4
CAS Number:
Molecular Weight:
231.53
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing