
L-Glutamic acid potassium salt monohydrate

Synonym(s):
(S)-2-Aminopentanedioic acid, L-α-Aminoglutaric acid potassium salt, L-2-Aminopentanedioic acid potassium salt, Glu, Potassium L-glutamate
Empirical Formula (Hill Notation):
C5H8KNO4 · H2O
CAS Number:
Molecular Weight:
203.23
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-2 of 2