
(-)-11-nor-9-Carboxy-delta8-THC solution

Synonym(s):
11-Nor-Δ8-tetrahydrocannabinol-9-carboxylic acid solution
Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing