
Guanosine 5′-triphosphate lithium salt

Synonym(s):
GTP lithium salt
Empirical Formula (Hill Notation):
C10H16N5O14P3 · xLi+
CAS Number:
Molecular Weight:
523.18 (free acid basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing